siRNA Details
| Gene Symbol: Atic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031014.2 | NP_112276.2 | 5 | 424 | ||
| GenBank mRNA | BC072496.1 | - | 5 | 424 | 7096880 | |
| D89514.1 | - | 5 | 417 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atic| Gene Symbol: Atic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031014.2 | NP_112276.2 | 5 | 424 | ||
| GenBank mRNA | BC072496.1 | - | 5 | 424 | 7096880 | |
| D89514.1 | - | 5 | 417 | |||