siRNA Details
| Gene Symbol: Chrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031016.1 | NP_112278.1 | 1 | 1125 | ||
| GenBank mRNA | AB017655.1 | - | 1 | 674 | ||
| J03025.1 | - | 1 | 1125 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrm2| Gene Symbol: Chrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031016.1 | NP_112278.1 | 1 | 1125 | ||
| GenBank mRNA | AB017655.1 | - | 1 | 674 | ||
| J03025.1 | - | 1 | 1125 | |||
Set Membership: |
|