siRNA Details
| Gene Symbol: Gabbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031028.3 | NP_112290.2 | 11 | 1354 | ||
| XM_006255879.2 | XP_006255941.1 | 11 | 1448 | |||
| XM_008772700.1 | XP_008770922.1 | 10 | 1427 | |||
| GenBank mRNA | AB016160.1 | - | 6 | 897 | ||
| AB016161.1 | - | 6 | 897 | |||
| AF283276.1 | - | 10 | 1268 | |||
| AJ831471.1 | - | 11 | 1352 | |||
| AJ831472.1 | - | 11 | 1476 | |||
| Y10369.2 | - | 11 | 1326 | |||
| Y10370.2 | - | 6 | 999 | |||
More Information
Set Membership: |
|

Gabbr1
Genomic Map