siRNA Details
| Gene Symbol: Mlh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031053.1 | NP_112315.1 | 2, 3 | 214 | ||
| GenBank mRNA | U80054.1 | - | 2, 3 | 214 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlh1| Gene Symbol: Mlh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031053.1 | NP_112315.1 | 2, 3 | 214 | ||
| GenBank mRNA | U80054.1 | - | 2, 3 | 214 | ||