siRNA Details
| Gene Symbol: Mvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031063.1 | NP_112325.1 | 6 | 629 | ||
| GenBank mRNA | M29472.1 | - | 6 | 629 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mvk| Gene Symbol: Mvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031063.1 | NP_112325.1 | 6 | 629 | ||
| GenBank mRNA | M29472.1 | - | 6 | 629 | ||