siRNA Details
| Gene Symbol: Bhmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030850.1 | NP_110477.1 | 4 | 516 | ||
| GenBank mRNA | AF038870.1 | - | 4 | 516 | ||
| BC078811.1 | - | 4 | 503 | 7130736 | ||
| U96133.1 | - | 4 | 505 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bhmt| Gene Symbol: Bhmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030850.1 | NP_110477.1 | 4 | 516 | ||
| GenBank mRNA | AF038870.1 | - | 4 | 516 | ||
| BC078811.1 | - | 4 | 503 | 7130736 | ||
| U96133.1 | - | 4 | 505 | |||