siRNA Details
| Gene Symbol: Mia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030852.2 | NP_110479.1 | 3 | 347 | ||
| GenBank mRNA | U51438.1 | - | 3 | 237 | ||
| U67884.1 | - | 3 | 347 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mia| Gene Symbol: Mia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030852.2 | NP_110479.1 | 3 | 347 | ||
| GenBank mRNA | U51438.1 | - | 3 | 237 | ||
| U67884.1 | - | 3 | 347 | |||