siRNA Details
| Gene Symbol: Lyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111098.1 | NP_001104568.1 | 4 | 434 | ||
| NM_030857.2 | NP_110484.1 | 4 | 497 | |||
| XM_006237834.2 | XP_006237896.1 | 4 | 813 | |||
| GenBank mRNA | AF000300.1 | - | 4 | 342 | ||
| AF000301.1 | - | 4 | 434 | |||
| AF000302.1 | - | 4 | 432 | |||
| L14782.1 | - | 3 | 291 | |||
| L14823.1 | - | 3 | 228 | |||
| L14951.1 | - | 4 | 500 | |||

Lyn
Genomic Map