siRNA Details
| Gene Symbol: Myoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030865.1 | NP_110492.1 | 1 | 499 | ||
| GenBank mRNA | AB019393.1 | - | 1 | 495 | ||
| AF093567.1 | - | 1 | 499 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myoc| Gene Symbol: Myoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030865.1 | NP_110492.1 | 1 | 499 | ||
| GenBank mRNA | AB019393.1 | - | 1 | 495 | ||
| AF093567.1 | - | 1 | 499 | |||