siRNA Details
| Gene Symbol: Ak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033967.2 | NP_001029139.1 | 5 | 551 | ||
| NM_030986.2 | NP_112248.2 | 5 | 551 | |||
| GenBank mRNA | BC061727.1 | - | 5 | 505 | 5598607 | |
| CO570724.1 | - | 5 | 551 | |||
| D13061.1 | - | 5 | 518 | |||
| FQ228915.1 | - | 3 | 212 | |||

Ak2
Genomic Map