siRNA Details
| Gene Symbol: Tg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270783.1 | NP_001257712.1 | 23 | 4773 | ||
| NM_030988.2 | NP_112250.2 | 24 | 4917 | |||
| GenBank mRNA | AB035201.1 | - | 24 | 4917 | ||
| AB679745.1 | - | 25 | 4891 | |||
| AB682738.1 | - | 24 | 4891 | |||
| AB682739.1 | - | 24 | 4747 | |||
| AF221622.1 | - | 24 | 4891 | |||

Tg
Genomic Map