siRNA Details
| Gene Symbol: Anpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031012.1 | NP_112274.1 | 12 | 1917 | ||
| XM_006229407.2 | XP_006229469.1 | 13 | 2337 | |||
| XM_006229408.2 | XP_006229470.1 | 13 | 2358 | |||
| XM_006229409.1 | XP_006229471.1 | 13 | 2394 | |||
| GenBank mRNA | M25073.1 | - | 12 | 1917 | ||
| M26710.1 | - | 12 | 1964 | |||
| Y18765.1 | - | 12 | 1307 | |||

Anpep
Genomic Map