siRNA Details
| Gene Symbol: Opn1sw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031015.1 | NP_112277.1 | 1, 2 | 402 | ||
| GenBank mRNA | AF051163.1 | - | 1, 2 | 354 | ||
| U63972.1 | - | 1, 2 | 402 | |||
More Information
Other Species Targeted
Set Membership: |
|

Opn1sw
Genomic Map