siRNA Details
| Gene Symbol: Ctbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031023.1 | NP_112285.1 | 4 | 532 | ||
| GenBank mRNA | BC088129.1 | - | 4 | 535 | 7323494 | |
| FQ219359.1 | - | 4 | 541 | |||
| FQ229296.1 | - | 2 | 155 | |||
| M95768.1 | - | 4 | 532 | |||

Ctbs
Genomic Map