siRNA Details
| Gene Symbol: Gatm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031031.2 | NP_112293.1 | 2, 3 | 352 | ||
| GenBank mRNA | BC081785.1 | - | 2, 3 | 352 | 7132560 | |
| U07971.1 | - | 2, 3 | 344 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gatm| Gene Symbol: Gatm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031031.2 | NP_112293.1 | 2, 3 | 352 | ||
| GenBank mRNA | BC081785.1 | - | 2, 3 | 352 | 7132560 | |
| U07971.1 | - | 2, 3 | 344 | |||