siRNA Details
| Gene Symbol: Gnaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031036.1 | NP_112298.1 | 6 | 848 | ||
| GenBank mRNA | AF234260.1 | - | 6 | 848 | ||
| DQ120477.1 | - | 6 | 813 | |||
| DQ120478.1 | - | 6 | 813 | |||
| L37294.1 | - | 1 | 119 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnaq| Gene Symbol: Gnaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031036.1 | NP_112298.1 | 6 | 848 | ||
| GenBank mRNA | AF234260.1 | - | 6 | 848 | ||
| DQ120477.1 | - | 6 | 813 | |||
| DQ120478.1 | - | 6 | 813 | |||
| L37294.1 | - | 1 | 119 | |||