siRNA Details
| Gene Symbol: Hnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031044.3 | NP_112306.1 | 3 | 254 | ||
| GenBank mRNA | BC087635.1 | - | 3 | 445 | 7319794 | |
| D10693.1 | - | 3 | 254 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnmt| Gene Symbol: Hnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031044.3 | NP_112306.1 | 3 | 254 | ||
| GenBank mRNA | BC087635.1 | - | 3 | 445 | 7319794 | |
| D10693.1 | - | 3 | 254 | |||