siRNA Details
| Gene Symbol: Jup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031047.2 | NP_112309.2 | 6 | 1358 | ||
| XM_006247429.1 | XP_006247491.1 | 6 | 1129 | |||
| XM_006247430.2 | XP_006247492.1 | 6 | 1095 | |||
| GenBank mRNA | BC065580.1 | - | 6 | 1358 | 5599724 | |
| FQ223166.1 | - | 6 | 1118 | |||
| U58858.1 | - | 6 | 1110 | |||

Jup
Genomic Map