siRNA Details
| Gene Symbol: Nell2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031070.2 | NP_112332.2 | 3 | 234 | ||
| XM_008765797.1 | XP_008764019.1 | 2 | 584 | |||
| XM_008765798.1 | XP_008764020.1 | 3 | 205 | |||
| GenBank mRNA | AY089719.1 | - | 3 | 234 | ||
| BC093617.1 | - | 2 | 178 | 7312010 | ||
| GQ376510.1 | - | 2 | 156 | |||
| U48245.1 | - | 3 | 234 | |||

Nell2
Genomic Map