siRNA Details
| Gene Symbol: Pggt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031082.1 | NP_112344.1 | 1, 2 | 157 | ||
| GenBank mRNA | L24116.1 | - | 1, 2 | 157 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pggt1b| Gene Symbol: Pggt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031082.1 | NP_112344.1 | 1, 2 | 157 | ||
| GenBank mRNA | L24116.1 | - | 1, 2 | 157 | ||