siRNA Details
| Gene Symbol: Grk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031096.1 | NP_112358.1 | 1 | 462 | ||
| GenBank mRNA | U63971.1 | - | 1 | 462 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grk1| Gene Symbol: Grk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031096.1 | NP_112358.1 | 1 | 462 | ||
| GenBank mRNA | U63971.1 | - | 1 | 462 | ||