siRNA Details
| Gene Symbol: St13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031122.1 | NP_112384.1 | 1 | 152 | ||
| GenBank mRNA | BC078804.1 | - | 1 | 159 | 7109473 | |
| FQ212434.1 | - | 209 | ||||
| FQ228746.1 | - | 1 | 114 | |||
| X82021.1 | - | 1 | 152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St13| Gene Symbol: St13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031122.1 | NP_112384.1 | 1 | 152 | ||
| GenBank mRNA | BC078804.1 | - | 1 | 159 | 7109473 | |
| FQ212434.1 | - | 209 | ||||
| FQ228746.1 | - | 1 | 114 | |||
| X82021.1 | - | 1 | 152 | |||