siRNA Details
| Gene Symbol: Cirbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031147.2 | NP_112409.2 | 2 | 249 | ||
| XM_006241012.2 | XP_006241074.1 | 2 | 254 | |||
| XM_006241013.2 | XP_006241075.2 | 2 | 365 | |||
| XM_006241014.2 | XP_006241076.1 | 2 | 254 | |||
| XM_006241015.2 | XP_006241077.2 | 2 | 365 | |||
| XM_008765132.1 | XP_008763354.1 | 2 | 365 | |||
| GenBank mRNA | AB000362.1 | - | 1 | 98 | ||
| BC069219.1 | - | 2 | 249 | 6888887 | ||

Cirbp
Genomic Map