siRNA Details
| Gene Symbol: Mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031151.2 | NP_112413.2 | 3 | 385 | ||
| GenBank mRNA | BC063165.1 | - | 3 | 385 | 6918633 | |
| DQ402951.1 | - | 2 | 219 | |||
| X04240.1 | - | 3 | 326 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mdh2| Gene Symbol: Mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031151.2 | NP_112413.2 | 3 | 385 | ||
| GenBank mRNA | BC063165.1 | - | 3 | 385 | 6918633 | |
| DQ402951.1 | - | 2 | 219 | |||
| X04240.1 | - | 3 | 326 | |||