siRNA Details
| Gene Symbol: Rab11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031152.2 | NP_112414.1 | 2 | 235 | ||
| GenBank mRNA | BC085727.1 | - | 2 | 235 | 7189729 | |
| FQ214239.1 | - | 2 | 299 | |||
| FQ220492.1 | - | 2 | 307 | |||
| FQ234527.1 | - | 2 | 237 | |||
| M75153.1 | - | 2 | 170 | |||

Rab11a
Genomic Map