siRNA Details
| Gene Symbol: Slit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031321.1 | NP_112611.1 | 2 | 523 | ||
| GenBank mRNA | AB011531.1 | - | 2 | 523 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slit3| Gene Symbol: Slit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031321.1 | NP_112611.1 | 2 | 523 | ||
| GenBank mRNA | AB011531.1 | - | 2 | 523 | ||