siRNA Details
| Gene Symbol: EPHB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004444.4 | NP_004435.3 | 14, 15 | 2980 | ||
| XM_017011816.1 | XP_016867305.1 | 14, 15 | 2637 | |||
| GenBank mRNA | AB209644.1 | - | 13, 14 | 3690 | ||
| AK301224.1 | - | 9, 10 | 1287 | |||
| AY056048.1 | - | 14, 15 | 2574 | |||
| BC004264.1 | - | 11, 12 | 2065 | |||
| BC052804.1 | - | 14, 15 | 2657 | 6584384 | ||
| U07695.1 | - | 14, 15 | 2574 | |||

EPHB4
Genomic Map