siRNA Details
| Gene Symbol: Nell1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031069.1 | NP_112331.1 | 5, 6 | 670 | ||
| GenBank mRNA | U48246.1 | - | 5, 6 | 670 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nell1| Gene Symbol: Nell1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031069.1 | NP_112331.1 | 5, 6 | 670 | ||
| GenBank mRNA | U48246.1 | - | 5, 6 | 670 | ||