siRNA Details
| Gene Symbol: Pde1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031078.2 | NP_112340.1 | 3, 4 | 657 | ||
| GenBank mRNA | L41045.1 | - | 3, 4 | 657 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde1c| Gene Symbol: Pde1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031078.2 | NP_112340.1 | 3, 4 | 657 | ||
| GenBank mRNA | L41045.1 | - | 3, 4 | 657 | ||