siRNA Details
| Gene Symbol: Soat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031118.1 | NP_112380.1 | 4 | 285 | ||
| XM_008769681.1 | XP_008767903.1 | 3 | 534 | |||
| GenBank mRNA | D86373.1 | - | 4 | 285 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Soat1| Gene Symbol: Soat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031118.1 | NP_112380.1 | 4 | 285 | ||
| XM_008769681.1 | XP_008767903.1 | 3 | 534 | |||
| GenBank mRNA | D86373.1 | - | 4 | 285 | ||