siRNA Details
| Gene Symbol: Vim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031140.1 | NP_112402.1 | 2, 3 | 712 | ||
| GenBank mRNA | BC061847.1 | - | 2, 3 | 752 | 5621065 | |
| FQ220610.1 | - | 1 | 15 | |||
| FQ229240.1 | - | 1 | 19 | |||
| FQ234991.1 | - | 2, 3 | 466 | |||
| X62952.1 | - | 2, 3 | 712 | |||

Vim
Genomic Map