siRNA Details
| Gene Symbol: Doc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031142.1 | NP_112404.1 | 4 | 778 | ||
| XM_008768117.1 | XP_008766339.1 | 4 | 839 | |||
| GenBank mRNA | U70778.2 | - | 4 | 778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Doc2b| Gene Symbol: Doc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031142.1 | NP_112404.1 | 4 | 778 | ||
| XM_008768117.1 | XP_008766339.1 | 4 | 839 | |||
| GenBank mRNA | U70778.2 | - | 4 | 778 | ||