siRNA Details
| Gene Symbol: Sh3gl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031238.2 | NP_112517.1 | 5 | 740 | ||
| XM_006229480.2 | XP_006229542.1 | 5 | 528 | |||
| XM_006229481.2 | XP_006229543.1 | 5 | 621 | |||
| XM_006229483.2 | XP_006229545.1 | 5 | 727 | |||
| XM_006229485.1 | XP_006229547.1 | 3 | 271 | |||
| XM_006229486.2 | XP_006229548.1 | 3 | 195 | |||
| GenBank mRNA | AB008159.2 | - | 2 | 185 | ||
| AF009604.1 | - | 3 | 224 | |||
| AF227439.1 | - | 5 | 743 | |||
| BC078800.1 | - | 5 | 743 | 7135194 | ||

Sh3gl3
Genomic Map