siRNA Details
Gene Symbol: Cyp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031241.1 | NP_112520.1 | 1 | 362 | ||
GenBank mRNA | AB009686.1 | - | 1 | 362 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cyp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031241.1 | NP_112520.1 | 1 | 362 | ||
GenBank mRNA | AB009686.1 | - | 1 | 362 |