siRNA Details
| Gene Symbol: Rock1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031098.1 | NP_112360.1 | 10 | 1243 | ||
| XM_006254400.2 | XP_006254462.1 | 10 | 1995 | |||
| XM_006254401.2 | XP_006254463.1 | 10 | 1995 | |||
| XM_008771957.1 | XP_008770179.1 | 10 | 1995 | |||
| GenBank mRNA | FQ225745.1 | - | 11 | 1719 | ||
| U61266.1 | - | 10 | 1243 | |||

Rock1
Genomic Map