siRNA Details
| Gene Symbol: Stc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031123.2 | NP_112385.1 | 3 | 603 | ||
| GenBank mRNA | BC078803.1 | - | 3 | 603 | 7123665 | |
| U62667.1 | - | 3 | 455 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stc1| Gene Symbol: Stc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031123.2 | NP_112385.1 | 3 | 603 | ||
| GenBank mRNA | BC078803.1 | - | 3 | 603 | 7123665 | |
| U62667.1 | - | 3 | 455 | |||