siRNA Details
| Gene Symbol: Tpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031137.1 | NP_112399.1 | 343 | |||
| XM_006244852.2 | XP_006244914.1 | 798 | ||||
| XM_006244853.2 | XP_006244915.1 | 798 | ||||
| XM_006244870.2 | XP_006244932.1 | 3 | 798 | |||
| GenBank mRNA | U50194.1 | - | 343 | |||

Tpp2
Genomic Map