siRNA Details
| Gene Symbol: Pax8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031141.2 | NP_112403.2 | 10, 11 | 1383 | ||
| XM_006233646.2 | XP_006233708.1 | 10, 11 | 2483 | |||
| XM_008761610.1 | XP_008759832.1 | 9, 10 | 2819 | |||
| XM_008761611.1 | XP_008759833.1 | 8, 9 | 2613 | |||
| XM_008761612.1 | XP_008759834.1 | 7, 8 | 2547 | |||
| XM_008761613.1 | XP_008759835.1 | 8, 9 | 2608 | |||
| GenBank mRNA | BC081779.1 | - | 10, 11 | 1383 | 7127710 | |
| FQ221890.1 | - | 2, 3 | 232 | |||
| X94246.1 | - | 9, 10 | 1266 | |||

Pax8
Genomic Map