siRNA Details
| Gene Symbol: Doc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031142.1 | NP_112404.1 | 1168 | |||
| GenBank mRNA | U70778.2 | - | 1168 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Doc2b| Gene Symbol: Doc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031142.1 | NP_112404.1 | 1168 | |||
| GenBank mRNA | U70778.2 | - | 1168 | |||