siRNA Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031143.1 | NP_112405.1 | 25 | 2423 | ||
| XM_006234594.2 | XP_006234656.1 | 26 | 3095 | |||
| XM_006234596.2 | XP_006234658.1 | 25 | 2258 | |||
| XM_008762037.1 | XP_008760259.1 | 25 | 4266 | |||
| GenBank mRNA | D78588.1 | - | 25 | 2423 | ||

Dgkz
Genomic Map