siRNA Details
| Gene Symbol: Arpc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031146.2 | NP_112408.1 | 6, 7 | 871 | ||
| GenBank mRNA | AF315378.1 | - | 5, 6 | 756 | ||
| CO396710.1 | - | 339 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arpc1a| Gene Symbol: Arpc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031146.2 | NP_112408.1 | 6, 7 | 871 | ||
| GenBank mRNA | AF315378.1 | - | 5, 6 | 756 | ||
| CO396710.1 | - | 339 | ||||