siRNA Details
| Gene Symbol: Fut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031236.1 | NP_112515.1 | 2 | 755 | ||
| XM_008759420.1 | XP_008757642.1 | 3 | 838 | |||
| GenBank mRNA | AB006137.1 | - | 2 | 755 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fut1| Gene Symbol: Fut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031236.1 | NP_112515.1 | 2 | 755 | ||
| XM_008759420.1 | XP_008757642.1 | 3 | 838 | |||
| GenBank mRNA | AB006137.1 | - | 2 | 755 | ||