siRNA Details
| Gene Symbol: Slc6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031343.1 | NP_112633.1 | 2, 3 | 456 | ||
| GenBank mRNA | AB021970.1 | - | 2, 3 | 456 | ||
| AB021971.1 | - | 2, 3 | 456 | |||
| Y13223.1 | - | 2, 3 | 414 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a2| Gene Symbol: Slc6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031343.1 | NP_112633.1 | 2, 3 | 456 | ||
| GenBank mRNA | AB021970.1 | - | 2, 3 | 456 | ||
| AB021971.1 | - | 2, 3 | 456 | |||
| Y13223.1 | - | 2, 3 | 414 | |||