siRNA Details
| Gene Symbol: Egfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031507.1 | NP_113695.1 | 1, 2 | 239 | ||
| GenBank mRNA | HM801041.1 | - | 1, 2 | 204 | ||
| HM801042.1 | - | 1, 2 | 204 | |||
| M37394.2 | - | 1, 2 | 239 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Egfr| Gene Symbol: Egfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031507.1 | NP_113695.1 | 1, 2 | 239 | ||
| GenBank mRNA | HM801041.1 | - | 1, 2 | 204 | ||
| HM801042.1 | - | 1, 2 | 204 | |||
| M37394.2 | - | 1, 2 | 239 | |||