siRNA Details
| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031517.2 | NP_113705.1 | 1 | 89 | ||
| XM_006236129.2 | XP_006236191.1 | 3 | 505 | |||
| GenBank mRNA | U65007.1 | - | 1 | 89 | ||
| X96786.1 | - | 1 | 89 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Met| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031517.2 | NP_113705.1 | 1 | 89 | ||
| XM_006236129.2 | XP_006236191.1 | 3 | 505 | |||
| GenBank mRNA | U65007.1 | - | 1 | 89 | ||
| X96786.1 | - | 1 | 89 | |||