siRNA Details
| Gene Symbol: Neu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031522.3 | NP_113710.2 | 1 | 310 | ||
| GenBank mRNA | AB035722.1 | - | 1 | 168 | ||
| BC100639.1 | - | 1 | 185 | 7929913 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Neu1| Gene Symbol: Neu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031522.3 | NP_113710.2 | 1 | 310 | ||
| GenBank mRNA | AB035722.1 | - | 1 | 168 | ||
| BC100639.1 | - | 1 | 185 | 7929913 | ||