siRNA Details
| Gene Symbol: Ampd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031544.1 | NP_113732.1 | 2 | 204 | ||
| XM_006230010.2 | XP_006230072.1 | 2 | 972 | |||
| XM_006230011.2 | XP_006230073.1 | 2 | 337 | |||
| XM_006230012.2 | XP_006230074.1 | 2 | 807 | |||
| XM_006230013.2 | XP_006230075.1 | 2 | 819 | |||
| XM_006230014.2 | XP_006230076.1 | 2 | 723 | |||
| XM_006230015.2 | XP_006230077.1 | 2 | 951 | |||
| XM_008759688.1 | XP_008757910.1 | 2 | 214 | |||
| GenBank mRNA | U90888.1 | - | 2 | 204 | ||

Ampd3
Genomic Map