siRNA Details
| Gene Symbol: Rgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031546.1 | NP_113734.1 | 2 | 175 | ||
| GenBank mRNA | AB037934.1 | - | 2 | 175 | ||
| BC078794.1 | - | 2 | 170 | 7126233 | ||
| D38467.1 | - | 2 | 169 | |||
| X69021.1 | - | 2 | 164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgn| Gene Symbol: Rgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031546.1 | NP_113734.1 | 2 | 175 | ||
| GenBank mRNA | AB037934.1 | - | 2 | 175 | ||
| BC078794.1 | - | 2 | 170 | 7126233 | ||
| D38467.1 | - | 2 | 169 | |||
| X69021.1 | - | 2 | 164 | |||