siRNA Details
| Gene Symbol: Scnn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031548.2 | NP_113736.1 | 1 | 457 | ||
| GenBank mRNA | AF082073.1 | - | 1 | 610 | ||
| BC070902.1 | - | 1 | 457 | 7100777 | ||
| EU627515.1 | - | 1 | 92 | |||
| U54699.1 | - | 1 | 124 | |||
| U54700.1 | - | 1 | 124 | |||
| X70497.1 | - | 1 | 170 | |||
| X70521.1 | - | 1 | 145 | |||

Scnn1a
Genomic Map