siRNA Details
| Gene Symbol: Smc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031583.2 | NP_113771.2 | 5 | 338 | ||
| GenBank mRNA | EV773232.1 | - | 2 | 102 | ||
| FQ231827.1 | - | 4 | 175 | |||
| FQ231919.1 | - | 5 | 350 | |||
| FQ233317.1 | - | 5 | 350 | |||
| FQ234437.1 | - | 5 | 350 | |||
| U82626.1 | - | 5 | 305 | |||

Smc3
Genomic Map